N-Boc-L-prolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(tert-Butoxycarbonyl)-L-prolinol, 1-Boc-2-(S)-pyrrolidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol, (S)-1-Boc-2-pyrrolidinemethanol, N-Boc-L-Prolinol

AS21921G100 AS21921G25
ACTIAS21921G100EA 0 GBP
ACTIAS21921G100 ACTIAS21921G25
N-Boc-L-prolinol
N-Boc-L-prolinol
Formula: C₁₀H₁₉NO₃
MDL Number: MFCD00066232
CAS Number: 69610-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR