4-Bromo-3-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21567G1 AS21567G5
ACTIAS21567G1EA 0 GBP
ACTIAS21567G1 ACTIAS21567G5
4-Bromo-3-hydroxypyridine
4-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
MDL Number: MFCD03093021
CAS Number: 161417-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR