4-Hydroxycyclohexanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Hydroxycyclohexanon

AS22161G1 AS22161G5
ACTIAS22161G1EA 0 GBP
ACTIAS22161G1 ACTIAS22161G5
4-Hydroxycyclohexanone
4-Hydroxycyclohexanone
Formula: C₆H₁₀O₂
MW: 114.1424 g/mol
MDL Number: MFCD02093817
CAS Number: 13482-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR