2-Hydroxy-3-methylpyrazine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Methylpyrazin-2-ol, 3-Methylpyrazin-2(1H)-one
AS21907G1
AS21907G5
ACTIAS21907G1EA
23
GBP
ACTIAS21907G1
ACTIAS21907G5
2-Hydroxy-3-methylpyrazine
2-Hydroxy-3-methylpyrazine
Formula:
C₅H₆N₂O MW: 110.11 g/mol |
MDL Number:
MFCD01646120 CAS Number: 19838-07-4 EINECS: 243-362-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...