tert-Butyl N-hydroxycarbamate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine
AS22433G100
AS22433G25
ACTIAS22433G100EA
0
GBP
ACTIAS22433G100
ACTIAS22433G25
tert-Butyl N-hydroxycarbamate
tert-Butyl N-hydroxycarbamate
Formula:
C₅H₁₁NO₃ MW: 133.15 g/mol |
MDL Number:
MFCD00002107 CAS Number: 36016-38-3 EINECS: 252-836-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...