1-Methylimidazole-4-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-1H-imidazole-4-carboxylic acid

AS20977G1 AS20977G5
ACTIAS20977G1EA 0 GBP
ACTIAS20977G1 ACTIAS20977G5
1-Methylimidazole-4-carboxylic acid
1-Methylimidazole-4-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD02179560
CAS Number: 41716-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR