1-Methylimidazole-2-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Methyl-1H-imidazole-2-carboxylic acid
AS21020G1
AS21020G25
AS21020G5
ACTIAS21020G1EA
19
GBP
ACTIAS21020G1
ACTIAS21020G25
ACTIAS21020G5
1-Methylimidazole-2-carboxylic acid
1-Methylimidazole-2-carboxylic acid
Formula:
C₅H₆N₂O₂ MW: 126.11 g/mol |
MDL Number:
MFCD01863421 CAS Number: 20485-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...