4-Iodotetrahydropyran

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Iodotetrahydro-2H-pyran

AS12602G1 AS12602G5
ACTIAS12602G1EA 0 GBP
ACTIAS12602G1 ACTIAS12602G5
4-Iodotetrahydropyran
4-Iodotetrahydropyran
Formula: C₅H₉IO
MDL Number: MFCD06797467
CAS Number: 25637-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR