Methyl 4-methoxyacetoacetate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23107G100 AS23107G25
ACTIAS23107G100EA 0 GBP
ACTIAS23107G100 ACTIAS23107G25
Methyl 4-methoxyacetoacetate
Methyl 4-methoxyacetoacetate
Formula: C₆H₁₀O₄
MDL Number: MFCD00010183
CAS Number: 41051-15-4
EINECS: 255-188-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR