1,5-Naphthyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22196G1 AS22196G5
ACTIAS22196G1EA 0 GBP
ACTIAS22196G1 ACTIAS22196G5
1,5-Naphthyridine
1,5-Naphthyridine
Formula: C₈H₆N₂
MDL Number: MFCD00039727
CAS Number: 254-79-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR