Bis(2,4-dimethoxybenzyl)amine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS34892G1
AS34892G5
ACTIAS34892G1EA
0
GBP
ACTIAS34892G1
ACTIAS34892G5
Bis(2,4-dimethoxybenzyl)amine
Bis(2,4-dimethoxybenzyl)amine
Formula:
C₁₈H₂₃NO₄ MW: 317.3800 g/mol |
MDL Number:
MFCD00040480 CAS Number: 20781-23-1 EINECS: 244-037-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...