N-Cbz-glycine(Z-Gly-OH)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

AS22061G100 AS22061G25 AS22061G5
ACTIAS22061G100EA 0 GBP
ACTIAS22061G100 ACTIAS22061G25 ACTIAS22061G5
N-Cbz-glycine(Z-Gly-OH)
N-Cbz-Glycine (Z-Gly-OH)
Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
MDL Number: MFCD00002691
CAS Number: 1138-80-3
EINECS: 214-516-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR