5-Bromopyrimidine-2-carbonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-cyanopyrimidine, 5-bromo-pyrimidine-2-carbonitile

AS20838G1 AS20838G5
ACTIAS20838G1EA 16 GBP
ACTIAS20838G1 ACTIAS20838G5
5-Bromopyrimidine-2-carbonitrile
5-Bromopyrimidine-2-carbonitrile
Formula: C₅H₂BrN₃
MW: 183.99 g/mol
MDL Number: MFCD02940446
CAS Number: 38275-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR