4,6-Dihydroxy-2-methylpyrimidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22367G100
AS22367G25
ACTIAS22367G100EA
0
GBP
ACTIAS22367G100
ACTIAS22367G25
4,6-Dihydroxy-2-methylpyrimidine
4,6-Dihydroxy-2-methylpyrimidine
Formula:
C₅H₆N₂O₂ |
MDL Number:
MFCD00006104 CAS Number: 40497-30-1 EINECS: 254-941-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...