2-Fluoro-4-hydroxybenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS31257G1 AS31257G5
ACTIAS31257G1EA 0 GBP
ACTIAS31257G1 ACTIAS31257G5
2-Fluoro-4-hydroxybenzaldehyde
2-Fluoro-4-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140.11 g/mol
MDL Number: MFCD06797918
CAS Number: 348-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR