9-Bromo-1-nonanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 9-Bromononan-1-ol

AS24439G100 AS24439G25
ACTIAS24439G100EA 89 GBP
ACTIAS24439G100 ACTIAS24439G25
9-Bromo-1-nonanol
9-Bromo-1-nonanol
Formula: C₉H₁₉BrO
MW: 223.15 g/mol
MDL Number: MFCD00063348
CAS Number: 55362-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR