AMPSO ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid

J62378.14 J62378.22
J62378.14EA 45.1 GBP
J62378.14 J62378.22
AMPSO ≥98%
AMPSO

Purity: 98+% CAS No.: 68399-79-1 Molecular Formula: C7H17NO5S Formula Weight: 227.28 Physical Form: Crystals Appearance: White Melting Point: 215-225° Beilstein Reference: 5929916 MDL No.: MFCD00041777


Useful as a buffer

Formula: C₇H₁₇NO₅S
MW: 227.28 g/mol
Melting Pt: 215…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00041777
CAS Number: 68399-79-1
EINECS: 269-991-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR