3-(Tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-(Tri-n-butylstannyl)pyridine

AS50319G25 AS50319G5
ACTIAS50319G25EA 0 GBP
ACTIAS50319G25 ACTIAS50319G5
3-(Tributylstannyl)pyridine
3-(Tributylstannyl)pyridine
Formula: C₁₇H₃₁NSn
MDL Number: MFCD00052054
CAS Number: 59020-10-9
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR