2-(Tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-(Tri-n-butylstannyl)pyridine

AS50318G100 AS50318G25 AS50318G5
ACTIAS50318G100EA 0 GBP
ACTIAS50318G100 ACTIAS50318G25 ACTIAS50318G5
2-(Tributylstannyl)pyridine
2-(Tributylstannyl)pyridine
Formula: C₁₇H₃₁NSn
MW: 368.15 g/mol
MDL Number: MFCD00052052
CAS Number: 17997-47-6
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR