3-Fluoro-2-(tributylstannyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS51256G1
AS51256G5
ACTIAS51256G1EA
0
GBP
ACTIAS51256G1
ACTIAS51256G5
3-Fluoro-2-(tributylstannyl)pyridine
3-Fluoro-2-(tributylstannyl)pyridine
Formula:
C₁₇H₃₀FNSn |
CAS Number:
573675-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...