(±)-Indoline-2-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Indoline-2-carboxylic acid
AS21192G100
AS21192G25
AS21192G5
ACTIAS21192G100EA
217
GBP
ACTIAS21192G100
ACTIAS21192G25
ACTIAS21192G5
(±)-Indoline-2-carboxylic acid
(±)-Indoline-2-carboxylic acid
Formula:
C₉H₉NO₂ MW: 163.18 g/mol |
MDL Number:
MFCD00010635 CAS Number: 78348-24-0 EINECS: 278-899-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...