1,3-Dichloroisoquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS51079G1 AS51079G5
ACTIAS51079G1EA 0 GBP
ACTIAS51079G1 ACTIAS51079G5
1,3-Dichloroisoquinoline
1,3-Dichloroisoquinoline
Formula: C₉H₅Cl₂N
MDL Number: MFCD00034750
CAS Number: 7742-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR