3-Bromophenyl phenyl ether (BDE No. 2)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromodiphenyl ether, 1-Bromo-3-phenoxybenzene

AS24221G25 AS24221G5
ACTIAS24221G25EA 449 GBP
ACTIAS24221G25 ACTIAS24221G5
3-Bromophenyl phenyl ether (BDE No. 2)
3-Bromophenyl phenyl ether (BDE No. 2)
Formula: C₁₂H₉BrO
MW: 249.1000 g/mol
MDL Number: MFCD00474520
CAS Number: 6876-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR