Glycine tert-butyl ester hydrochloride (H-Gly-OtBu.HCl)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

AS21095G100 AS21095G25
ACTIAS21095G100EA 0 GBP
ACTIAS21095G100 ACTIAS21095G25
Glycine tert-butyl ester hydrochloride (H-Gly-OtBu.HCl)
Glycine tert-butyl ester hydrochloride (H-Gly-OtBu.HCl)
Formula: C₆H₁₄ClNO₂
MW: 167.64 g/mol
MDL Number: MFCD00058255
CAS Number: 27532-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR