Pyrrole-3-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1H-Pyrrole-3-carboxylic acid

AS21396G1 AS21396G25 AS21396G5
ACTIAS21396G1EA 39 GBP
ACTIAS21396G1 ACTIAS21396G25 ACTIAS21396G5
Pyrrole-3-carboxylic acid
Pyrrole-3-carboxylic acid
Formula: C₅H₅NO₂
MW: 111.10 g/mol
MDL Number: MFCD00800594
CAS Number: 931-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR