5-Bromoindoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25055G25 AS25055G5
ACTIAS25055G25EA 0 GBP
ACTIAS25055G25 ACTIAS25055G5
5-Bromoindoline
5-Bromoindoline
Formula: C₈H₈BrN
MW: 198.06 g/mol
MDL Number: MFCD00027410
CAS Number: 22190-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR