1-(2-Methoxy-4-nitrophenyl)pyrrolidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20475G1
AS20475G5
ACTIAS20475G1EA
0
GBP
ACTIAS20475G1
ACTIAS20475G5
1-(2-Methoxy-4-nitrophenyl)pyrrolidine
1-(2-Methoxy-4-nitrophenyl)pyrrolidine
Formula:
C₁₁H₁₄N₂O₃ |
MDL Number:
MFCD00096778 CAS Number: 67828-57-3 EINECS: 267-278-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...