1-Bromoisoquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS31177G1 AS31177G5
ACTIAS31177G1EA 0 GBP
ACTIAS31177G1 ACTIAS31177G5
1-Bromoisoquinoline
1-Bromoisoquinoline
Formula: C₉H₆BrN
MDL Number: MFCD00234478
CAS Number: 1532-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR