3-Bromo-5-fluoroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22740G100 AS22740G25 AS22740G5
ACTIAS22740G100EA 0 GBP
ACTIAS22740G100 ACTIAS22740G25 ACTIAS22740G5
3-Bromo-5-fluoroanisole
3-Bromo-5-fluoroanisole
Formula: C₇H₆BrFO
MDL Number: MFCD01861129
CAS Number: 29578-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR