2-Chloro-5-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20543G100 AS20543G25
ACTIAS20543G100EA 0 GBP
ACTIAS20543G100 ACTIAS20543G25
2-Chloro-5-fluorobenzaldehyde
2-Chloro-5-fluorobenzaldehyde
Formula: C₇H₄ClFO
MDL Number: MFCD03788511
CAS Number: 84194-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR