2,4,5-T-(2,4,5-Trichlorophenoxy acetic acid)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23055G1 AS23055G5
ACTIAS23055G1EA 99 GBP
ACTIAS23055G1 ACTIAS23055G5
2,4,5-T-(2,4,5-Trichlorophenoxy acetic acid)
2,4,5-T (2,4,5-trichlorophenoxy acetic acid)
Formula: C₈H₅Cl₃O₃
MW: 255.48 g/mol
MDL Number: MFCD00004301
CAS Number: 93-76-5
EINECS: 202-273-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR