Diethyl isopropylidenemalonate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22291G100 AS22291G25
ACTIAS22291G100EA 0 GBP
ACTIAS22291G100 ACTIAS22291G25
Diethyl isopropylidenemalonate
Diethyl isopropylidenemalonate
Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
MDL Number: MFCD00009147
CAS Number: 6802-75-1
EINECS: 229-876-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR