2-Amino-1,3,4-thiadiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

AS20907G100 AS20907G25
ACTIAS20907G100EA 167 GBP
ACTIAS20907G100 ACTIAS20907G25
2-Amino-1,3,4-thiadiazole
2-Amino-1,3,4-thiadiazole
Formula: C₂H₃N₃S
MW: 101.13 g/mol
MDL Number: MFCD00003107
CAS Number: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR