2-(2-Bromoethyl)-1,3-dioxolane

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22276G100 AS22276G25
ACTIAS22276G100EA 0 GBP
ACTIAS22276G100 ACTIAS22276G25
2-(2-Bromoethyl)-1,3-dioxolane
2-(2-Bromoethyl)-1,3-dioxolane
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
MDL Number: MFCD00003216
CAS Number: 18742-02-4
EINECS: 242-551-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR