2-Amino-4-chloro-6-methylpyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23246G100 AS23246G25
ACTIAS23246G100EA 0 GBP
ACTIAS23246G100 ACTIAS23246G25
2-Amino-4-chloro-6-methylpyrimidine
2-Amino-4-chloro-6-methylpyrimidine
Formula: C₅H₆ClN₃
MDL Number: MFCD00006091
CAS Number: 5600-21-5
EINECS: 227-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR