Ethyl isobutyrylacetate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 4-methyl-3-oxopentanoate

AS24057G100 AS24057G25
ACTIAS24057G100EA 168 GBP
ACTIAS24057G100 ACTIAS24057G25
Ethyl isobutyrylacetate
Ethyl isobutyrylacetate
Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD00009198
CAS Number: 7152-15-0
EINECS: 230-491-9
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR