4-Piperidinemethanol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Piperidin-4-ylmethanol

AS20182G100 AS20182G25 AS20182G5
ACTIAS20182G100EA 0 GBP
ACTIAS20182G100 ACTIAS20182G25 ACTIAS20182G5
4-Piperidinemethanol
4-Piperidinemethanol
Formula: C₆H₁₃NO
MDL Number: MFCD00174228
CAS Number: 6457-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR