4,6-Dichloro-5-nitropyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20001G100 AS20001G25
ACTIAS20001G100EA 0 GBP
ACTIAS20001G100 ACTIAS20001G25
4,6-Dichloro-5-nitropyrimidine
4,6-Dichloro-5-nitropyrimidine
Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
MDL Number: MFCD00006107
CAS Number: 4316-93-2
EINECS: 224-340-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR