4-Guanidinobenzoic acid hydrochloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21994G100 AS21994G25
ACTIAS21994G100EA 0 GBP
ACTIAS21994G100 ACTIAS21994G25
4-Guanidinobenzoic acid hydrochloride
4-Guanidinobenzoic acid hydrochloride
Formula: C₈H₁₀ClN₃O₂
MDL Number: MFCD00040587
CAS Number: 42823-46-1
EINECS: 255-956-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR