Benzyl carbazate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate

AS24943G100 AS24943G25
ACTIAS24943G100EA 0 GBP
ACTIAS24943G100 ACTIAS24943G25
Benzyl carbazate
Benzyl carbazate
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00041890
CAS Number: 5331-43-1
EINECS: 226-230-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR