3-(4-Bromobenzoyl)propionic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23378G100 AS23378G25 AS23378G5
ACTIAS23378G100EA 0 GBP
ACTIAS23378G100 ACTIAS23378G25 ACTIAS23378G5
3-(4-Bromobenzoyl)propionic acid
3-(4-Bromobenzoyl)propionic acid
Formula: C₁₀H₉BrO₃
MDL Number: MFCD00016563
CAS Number: 6340-79-0
EINECS: 228-732-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR