3,4-Dibromothiophene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20077G100 AS20077G25
ACTIAS20077G100EA 0 GBP
ACTIAS20077G100 ACTIAS20077G25
3,4-Dibromothiophene
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 4.5 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR