3-(Trifluoromethyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21477G25 AS21477G5
ACTIAS21477G25EA 0 GBP
ACTIAS21477G25 ACTIAS21477G5
3-(Trifluoromethyl)pyridine
3-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
Boiling Pt: 110…113 °C (1013 hPa)
MDL Number: MFCD00100032
CAS Number: 3796-23-4
EINECS: 223-268-2
UN: 1992
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR