2-Bromo-5-methylpyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20026G100
AS20026G25
ACTIAS20026G100EA
0
GBP
ACTIAS20026G100
ACTIAS20026G25
2-Bromo-5-methylpyridine
2-Bromo-5-methylpyridine
Formula:
C₆H₆BrN Boiling Pt: 65 °C (0,5 mmHg) Melting Pt: 152,5 °C |
MDL Number:
MFCD00209553 CAS Number: 3510-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...