4-Methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20716G25 AS20716G5
ACTIAS20716G25EA 0 GBP
ACTIAS20716G25 ACTIAS20716G5
4-Methoxypyridine
4-Methoxypyridine
Formula: C₆H₇NO
MDL Number: MFCD00674049
CAS Number: 620-08-6
EINECS: 210-624-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR