2-Fluoro-5-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23161G1 AS23161G25 AS23161G5
ACTIAS23161G1EA 0 GBP
ACTIAS23161G1 ACTIAS23161G25 ACTIAS23161G5
2-Fluoro-5-nitropyridine
2-Fluoro-5-nitropyridine
Formula: C₅H₃FN₂O₂
MDL Number: MFCD03095059
CAS Number: 456-24-6
EINECS: 207-260-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR