3-Bromo-2-chloropyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS21322G100
AS21322G25
ACTIAS21322G100EA
0
GBP
ACTIAS21322G100
ACTIAS21322G25
3-Bromo-2-chloropyridine
3-Bromo-2-chloropyridine
Formula:
C₅H₃BrClN Boiling Pt: 205 °C (1013 hPa) Melting Pt: 55 °C |
MDL Number:
MFCD00234007 CAS Number: 52200-48-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...