2-(Bromomethyl)naphthalene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24437G100
AS24437G25
ACTIAS24437G100EA
0
GBP
ACTIAS24437G100
ACTIAS24437G25
2-(Bromomethyl)naphthalene
2-(Bromomethyl)naphthalene
Formula:
C₁₁H₉Br Boiling Pt: 293 °C (1013 hPa) Melting Pt: 60…62 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004123 CAS Number: 939-26-4 EINECS: 213-359-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...