1-Isoquinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Isocarbostyril, 1-Hydroxyisoquinoline

AS21176G25 AS21176G5
ACTIAS21176G25EA 0 GBP
ACTIAS21176G25 ACTIAS21176G5
1-Isoquinolinol
1-Isoquinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006899
CAS Number: 491-30-5
EINECS: 207-732-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR