2-Indanone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one

AS20096G100 AS20096G25 AS20096G5
ACTIAS20096G100EA 0 GBP
ACTIAS20096G100 ACTIAS20096G25 ACTIAS20096G5
2-Indanone
2-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 220…225 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.0712 g/cm³ (67 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003792
CAS Number: 615-13-4
EINECS: 210-410-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR